missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Lactide, TRC

Product Code. 30477865
Change view
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477865 250 g 250g
30465349 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477865 Supplier Toronto Research Chemicals Supplier No. TRCL113600250G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4511-42-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
Synonym (3S,6S)-3,6-Dimethyl-1,4-dioxane-2,5-dione, (3S-cis)-3,6-Dimethyl-1,4-dioxane-2,5-dione, L-3,6-Dimethyl-p-dioxane-2,5-dione, (-)-L-Dilactide, (S,S)-3,6-Dimethyl-1,4-dioxane-2,5-dione, L,L-Dilactide, L-Dilactide, L-Lactide, LL-Lactide, Purasorb L
IUPAC Name (3S,6S)-3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES C[C@@H]1OC(=O)[C@H](C)OC1=O

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H8O4/c1-3-5(7)10-4(2)6(8)9-3/h3-4H,1-2H3/t3-,4-/m0/s1
Formula Weight 144.0423
Chemical Name or Material L-(-)-Lactide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.