missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, 98.5 to 101.5% (Dry Basis), Ultrapure

Product Code. 15875168 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15875168 100 g Plastic Bottle 100g
15885308 1 kg Plastic Bottle 1kg
15895308 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15875168 Supplier Thermo Scientific Supplier No. J18280.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 61-90-5 | C6H13NO2 | 131.18 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2 S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Melting Point >300°C
Packaging Plastic Bottle
Beilstein 1721722
Density 1.293
Quantity 100 g
Formula Weight 131.17
Percent Purity 98.5 to 101.5%
Specific Gravity 1.293
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Leucine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.