missing translation for 'onlineSavingsMsg'
Learn More

L-leucine, 99.42%, MP Biomedicals™

Product Code. 11319019 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319019 25 g 1kg
11329019 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319019 Supplier MP Biomedicals Supplier No. 0210215801
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 0.9942
Density 1.293g/mL
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
Quantity 25 g
Formula Weight 131.18
Physical Form Small Lustrous Plates or Crystalline Powder
Chemical Name or Material L-Leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.