missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine-d10-N-FMOC, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine-2,3,3,4,5,5,5,5',5',5'-D10 |
| CAS | 1190594-22-9 |
| Molecular Formula | C21 D10 H13 N O4 |
| Quantity | 0.25 g |
| Synonym | L-Leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-[(9 H-fluoren-9-ylmethoxy)carbonyl]- (ACI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine-2,3,3,4,5,5,5,5',5',5'-d10 (ACI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-leucine-2,3,3,4,5,5,5,5',5',5'-D10, L-Leucine-d10-N-FMOC, (2 S)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-(methyl-d3)pentanoic-2,3,3,4,5,5,5-d7 acid, N-FMOC-L-leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-FMOC-L-leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-FMOC-L-leucine-2,3,3,4,5,5,5,5',5',5'-d10 |
| InChI Formula | InChI=1 S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19 H,11-12H2,1-2H3,(H,22,25)(H,23,24)/t19-/m0/s1/i1D3,2D3,11D2,13 D,19 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[C@]([2 H])(NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O |
| IUPAC Name | (2 S)-2,3,3,4,5,5,5-heptadeuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-4-(trideuteriomethyl)pentanoic acid |
| Molecular Weight (g/mol) | 363.4732 |
| Formula Weight | 363.2255 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction