missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15547964 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15547964 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15547964 Supplier Avantor J.T.Baker Supplier No. 208306

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 61-90-5
Molecular Formula C6H13NO2
Packaging Poly Bottle
MDL Number MFCD00002617
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
SMILES CC(C)C[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
Molecular Weight (g/mol) 131.18
Quantity 1 kg
PubChem CID 6106
ChEBI CHEBI:15603
Formula Weight 131.17
Specific Gravity 1L = 1.29kg
Grade Multi-Compendial/USP
Chemical Name or Material L-Leucine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.