missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10637823
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10637823 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10637823 Supplier Fisher BioReagents Supplier No. BP385100
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 286°C
Ammonium 0.02% max.
CAS 61-90-5
Arsenic (As) 1ppm max.
pH 5.5 to 6.5
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.2% max.
Other Amino Acids Chromatographically not detectable
Molecular Formula C6H13NO2
Packaging Glass Bottle
MDL Number MFCD00002617
Specific Rotation 14.9 to 16.1° (+ or -)
Merck Index 15, 5503
State of Solution (%T) 98% min.
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
Alpha Vector Leucine
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
SMILES CC(C)C[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
Molecular Weight (g/mol) 131.18
Quantity 100 g
PubChem CID 6106
ChEBI CHEBI:15603
Formula Weight 131.17
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Leucine
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.