missing translation for 'onlineSavingsMsg'
Learn More

L-Leucinol, 97%

Product Code. 11420178
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11420178 5 g 5g
11430178 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11420178

Brand: Thermo Scientific Alfa Aesar B23745.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7533-40-6 | C6H15NO | 117.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7533-40-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00063676
InChI Key VPSSPAXIFBTOHY-LURJTMIESA-N
Synonym l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol
PubChem CID 111307
IUPAC Name (2S)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@H](N)CO

Specifications

Refractive Index 1.451
Density 0.917
Boiling Point 198°C to 200°C
Quantity 5 g
Flash Point 90°C (194°F)
Optical Rotation +1.1° (Neat)
Sensitivity Air Sensitive
Beilstein 1719240
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material L-Leucinol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.