missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine-2,3,3,4,4,5,5,6,6-d9 HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | L-Lysine-D9 Monohydrochloride |
| CAS | 2708343-64-8 |
| Molecular Formula | C6 D9 H5 N2 O2 . Cl H |
| Quantity | 0.1 g |
| Synonym | L-Lysine-d9 hydrochloride, L-Lysine-2,3,3,4,4,5,5,6,6-d9 hydrochloride (1:1), L-Lysine-D9 monohydrochloride, (2 S)-2,6-Diaminohexanoic-2,3,3,4,4,5,5,6,6-d9 acid monohydrochloride, (2 S)-2,6-Diamino-2,3,3,4,4,5,5,6,6-nonadeuteriohexanoic acid monohydrochloride |
| InChI Formula | InChI=1 S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1 H/t5-;/m0./s1/i1D2,2D2,3D2,4D2,5 D; |
| SMILES | Cl.[2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2,6-diamino-2,3,3,4,4,5,5,6,6-nonadeuteriohexanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 191.704 |
| Formula Weight | 191.1387 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction