missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine, 98%

Product Code. 15197412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15197412 500 g Glass Bottle 500g
10016203 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15197412 Supplier Thermo Scientific Chemicals Supplier No. 303345000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 56-87-1 | C6H14N2O2 | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-87-1
Molecular Formula C6H14N2O2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00064433
InChI Key KDXKERNSBIXSRK-MLHKIVSYNA-N
Synonym l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum
PubChem CID 5962
ChEBI CHEBI:18019
IUPAC Name (2S)-2,6-diaminohexanoic acid
SMILES NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 214.0°C to 216.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation +23.0° to +27.0° (20°C, 589nm) (C=2, 1N HCl)
Water 3% max.
Quantity 500 g
Formula Weight 146.19
Percent Purity 98%
Physical Form Powder or Crystals
Chemical Name or Material L-Lysine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.