missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Lysine Acetate

Product Code. 30018435
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018435 100 g 100g
30018436 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018435 Supplier Melford Supplier No. L37020100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Lysine Acetate is employed in biotechnological processes, especially in the production of recombinant proteins and enzymes.

L-Lysine Acetate is employed in biotechnological processes, especially in the production of recombinant proteins and enzymes. It plays a role in cell culture media to support the growth and productivity of cells.

Additionally, L-Lysine Acetate is utilized in laboratory research, particularly in studies related to protein synthesis, enzymology, and other biochemical processes.

Its role as a fundamental amino acid makes it valuable in understanding various cellular and molecular mechanisms.

Specifications

Chemical Name or Material L-Lysine Acetate
CAS 57282-49-2
Molecular Formula C8H18N2O4
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.