missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine acetate, 97%

Product Code. 15294874 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15294874 25 g 25g
15214884 100 g 100g
15224884 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15294874 Supplier Thermo Scientific Acros Supplier No. 451630250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57282-49-2 | C8H18N2O4 | 206.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57282-49-2
Molecular Formula C8H18N2O4
Molecular Weight (g/mol) 206.24
MDL Number MFCD00039069
InChI Key RRNJROHIFSLGRA-JEDNCBNOSA-N
Synonym l-lysine acetate, l-lysine acetate salt, lysine acetate, l-lysine monoacetate, unii-ttl6g7liwz, lysine acetate salt, lysine, monoacetate, lysine acetate usp, l-lysine, acetate, ttl6g7liwz
PubChem CID 104152
IUPAC Name acetic acid;(2S)-2,6-diaminohexanoic acid
SMILES CC(O)=O.NCCCC[C@H](N)C(O)=O

Specifications

Assay Percent Range 97%
Quantity 25 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material L-Lysine acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.