missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine acetate, TRC

Product Code. 30478545
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478545 10 g 10g
30472072 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478545 Supplier Toronto Research Chemicals Supplier No. TRCL48877810G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lysine acetate
CAS 57282-49-2
Purity Grade Notes HPLC
Molecular Formula C6 H14 N2 O2 . C2 H4 O2
Quantity 10 g
Synonym L-Lysine, acetate (1:1), -Lysine, monoacetate (9CI), L-Lysine acetate, Lysine monoacetate, Lysine acetate, (2S)-2,6-Diaminohexanoic acid acetate
InChI Formula InChI=1S/C6H14N2O2.C2H4O2/c7-4-2-1-3-5(8)6(9)10;1-2(3)4/h5H,1-4,7-8H2,(H,9,10);1H3,(H,3,4)/t5-;/m0./s1
SMILES CC(=O)O.NCCCC[C@H](N)C(=O)O
IUPAC Name acetic acid;(2S)-2,6-diaminohexanoic acid
Molecular Weight (g/mol) 206.24
Formula Weight 206.1267
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.