missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine hydrochloride, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals

Product Code. 15805318 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Packaging:
Fiber Drum
Plastic Bottle
Poly Pail
Unit Size:
10kg
1kg
5kg
Product Code. Quantity Packaging unitSize
15805318 1 kg Plastic Bottle 1kg
15815318 5 kg Poly Pail 5kg
15825318 10 kg Fiber Drum 10kg
3 options
This item is not returnable. View return policy

Product Code. 15805318

Brand: Thermo Scientific J18585.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.648
MDL Number MFCD00064564
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
SMILES C(CCN)CC(C(=O)O)N.Cl

Specifications

Melting Point ∼260°C (decomposition)
Packaging Plastic Bottle
Linear Formula NH2(CH2)4CH(NH2)COOH.HCl
Beilstein 3563889
IUPAC Name (2 S)-2,6-diaminohexanoic acid;hydrochloride
Quantity 1 kg
Formula Weight 182.65
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Lysine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.