missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine hydrochloride, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals

Product Code. 15825318 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
10 kg
Packaging:
Fiber Drum
Plastic Bottle
Poly Pail
Unit Size:
10kg
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15825318 10 kg Fiber Drum 10kg
15805318 1 kg Plastic Bottle 1kg
15815318 5 kg Poly Pail 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15825318 Supplier Thermo Scientific Supplier No. J18585.A9
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 657-27-2 | Molecular Formula: C6H15ClN2O2 | Molecular Weight: 182.65 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.648
MDL Number MFCD00064564
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
SMILES C(CCN)CC(C(=O)O)N.Cl

Specifications

Melting Point ∼260°C (decomposition)
Packaging Fiber Drum
Linear Formula NH2(CH2)4CH(NH2)COOH.HCl
Beilstein 3563889
IUPAC Name (2 S)-2,6-diaminohexanoic acid;hydrochloride
Quantity 10 kg
Formula Weight 182.65
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Lysine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.