missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Hydrochloride, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15557964 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15557964 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557964 Supplier Avantor J.T.Baker Supplier No. 208406

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Packaging Poly Bottle
MDL Number MFCD00064564 MFCD00081870
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
SMILES C(CCN)CC(C(=O)O)N.Cl
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
Molecular Weight (g/mol) 182.648
Quantity 1 kg
PubChem CID 69568
ChEBI CHEBI:53633
Formula Weight 182.65
Grade Multi-Compendial/USP
Chemical Name or Material L-Lysine Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.