missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Lysine Monohydrate

Product Code. 30018438
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018438 500 g 500g
30018437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018438 Supplier Melford Supplier No. L37050500.0
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • CAS Number: 39665-12-8
  • Molecular Formula: C6H14N2O2 H2O
  • Molecular Weight: 164.21g/mol

Specifications

Chemical Name or Material L-Lysine Monohydrate
CAS 39665-12-8
Molecular Formula C6H14N2O2-H2O
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.