missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine monohydrate, 97%

Product Code. 11499963
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499963 25 g 25g
11409973 100 g 100g
11419973 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499963 Supplier Thermo Scientific Alfa Aesar Supplier No. A13040.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39665-12-8 | C6H16N2O3 | 164.21 g/mol

L-Lysine is an essential proteinogenic alfa amino acid used in various applications such as non-animal sourced supplement in cell culture media. It acts as a substrate of enzymes like L-lysine oxidase as well as a component of poly-lysine polymers. It is also involved in the studies of glycation mechanism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine is an essential proteinogenic alfa amino acid used in various applications such as non-animal sourced supplement in cell culture media. It acts as a substrate of enzymes like L-lysine oxidase as well as a component of poly-lysine polymers. It is also involved in the studies of glycation mechanism.

Solubility
Soluble in water and hydrochloric acid.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
SMILES O.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 210°C (decomposition)
Beilstein 1722531
Merck Index 14,5636
Solubility Information Soluble in water and hydrochloric acid.
IUPAC Name 2,6-diaminohexanoic acid hydrate
Quantity 25 g
Formula Weight 164.21 (146.19 Anhydrous)
Optical Rotation +22.5° (c=2 in 6M HCl)
Sensitivity Air sensitive; Hygroscopic
Percent Purity 97%
Chemical Name or Material L-Lysine monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.