missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Monohydrochloride, Extra Pure, Fisher Chemical™

Product Code. 10764951 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
Product Code. Quantity unitSize
10764951 100 g 100g
11478972 1 kg 1kg
2 options
This item is not returnable. View return policy

Product Code. 10764951

Brand: Fisher Chemical L/2700/48

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.648
MDL Number 64564
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
SMILES C(CCN)CC(C(=O)O)N.Cl

Specifications

Melting Point 263°C
Color White
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 182.65g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.