missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine, TRC

Product Code. 30137374
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137374 1 g 1g
30005911 10 mg 10mg
30009441 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137374 Supplier Toronto Research Chemicals Supplier No. TRCL4688951G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-87-1
Molecular Formula C6 H14 N2 O2
Molecular Weight (g/mol) 146.19
Synonym L-Lysine, Lysine, L- (8CI), (+)-S-Lysine, (S)-2,6-Diaminohexanoic acid, (S)-Lysine, (S)-α,ε-Diaminocaproic acid, 2,6-Diaminohexanoic acid, Aminutrin, Biolys, Hexanoic acid, 2,6-diamino-, (S)-, L-(+)-Lysine, L-2,6-Diaminocaproic acid, L-Lys, L-Norleucine, 6-amino-, LLB 50, Lysine, Lysine acid, Malandil, h-Lys-oh, α-Lysine
IUPAC Name (2S)-2,6-diaminohexanoic acid
SMILES NCCCC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C6H14N2O2/c7-4-2-1-3-5(8)6(9)10/h5H,1-4,7-8H2,(H,9,10)/t5-/m0/s1
Quantity 1 g
Formula Weight 146.1055
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-(+)-Lysine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.