missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Malic Acid, TRC

Product Code. 30137870
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137870 25 g 25g
30136299 100 g 100g
17980895 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137870 Supplier Toronto Research Chemicals Supplier No. TRCM15950525G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-67-6
Molecular Formula C4 H6 O5
Molecular Weight (g/mol) 134.09
Synonym L-Malic Acid, (2S)-2-Hydroxybutanedioic acid, Butanedioic acid, hydroxy-, (2S)- (9CI), Butanedioic acid, hydroxy-, (S)-, Malic acid, L- (8CI), Malic acid, l- (3CI), (-)-(S)-Malic acid, (-)-Hydroxysuccinic acid, (-)-L-Malic acid, (-)-Malic acid, (2S)-2-Hydroxybutanedioic acid, (2S)-2-Hydroxysuccinic acid, (2S)-Malic acid, (S)-2-Hydroxysuccinic acid, (S)-Malic acid, Apple acid, L-(-)-Malic acid, L-Apple acid, L-Malic acid, NSC 9232, S-(-)-Malic acid, S-2-Hydroxybutanedioic acid
IUPAC Name (2S)-2-hydroxybutanedioic acid
SMILES O[C@@H](CC(=O)O)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m0/s1
Formula Weight 134.0215
Chemical Name or Material L-(-)-Malic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.