missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Mannose, 99+%, Thermo Scientific Chemicals

Product Code. 10665842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665842 1 g Glass bottle 1g
10184710 250 mg Glass bottle 250mg
10464155 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10665842 Supplier Thermo Scientific Acros Supplier No. 227590010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10030-80-5 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10030-80-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00136021
InChI Key GZCGUPFRVQAUEE-BXKVDMCESA-N
Synonym l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
PubChem CID 82308
ChEBI CHEBI:37681
IUPAC Name (2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O

Specifications

Melting Point 129.0°C to 131.0°C
Color White
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (60° to 70°C)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition −14.70° (20°C c=4,H2O,NH3)
Specific Rotation − 14.70
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,insoluble in ether
Formula Weight 180.16
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material L(-)-Mannose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.