missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Mannose, 99%

Product Code. 11413318
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413318 0.25 g 0.25g
11403318 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413318 Supplier Thermo Scientific Chemicals Supplier No. A17722.01
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10030-80-5 | C6H12O6 | 180.16 g/mol

L-Mannose is used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Mannose is used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.

Solubility
Soluble in Water (100 mg/mL).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10030-80-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00136021
InChI Key GZCGUPFRVQAUEE-BXKVDMCESA-N
Synonym l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
PubChem CID 82308
ChEBI CHEBI:37681
IUPAC Name (2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O

Specifications

Melting Point 129°C to 131°C
Quantity 0.25 g
Beilstein 1724628
Solubility Information Soluble in Water (100mg/mL).
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material L-(-)-Mannose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.