missing translation for 'onlineSavingsMsg'
Learn More

l-Menthone, 85%

Product Code. 10792051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10792051 100 mL Glass bottle 100mL
10308250 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10792051 Supplier Thermo Scientific Acros Supplier No. 203611000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14073-97-3 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14073-97-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001634,MFCD00136033
InChI Key NFLGAXVYCFJBMK-BDAKNGLRSA-N
Synonym --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
PubChem CID 26447
ChEBI CHEBI:15410
IUPAC Name (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one
SMILES CC(C)[C@@H]1CC[C@@H](C)CC1=O

Specifications

Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 207°C to 210°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.453
Quantity 100 mL
Beilstein 07, 38
Fieser 14,201; 16,204
Merck Index 15, 5906
Specific Gravity 0.89
Specific Rotation Condition −6° (20°C c=4,ethanol)
Specific Rotation -6
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,miscible with ethanol and,ethyl ether
Formula Weight 154.25
Percent Purity 85%
Physical Form Liquid
Chemical Name or Material l-Menthone, 85%
Show More Show Less
compliance-icons
Product Identifier
  • l-Menthone
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.