missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, 98+%

Product Code. 11434083
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434083 500 g 500g
11424083 100 g 100g
11444083 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434083 Supplier Thermo Scientific Chemicals Supplier No. A10318.36
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-Methionine (CAS # 63-68-3) is an essential amino acid that contains sulphur. L-Methionine is involved in cellular detoxification in several species. | CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol

L-Methionine iserves as a chelating agent for heavy metals, as a flavoring agent and nutritional supplement for foods. It acts as a feed additive, vegetable oil enrichment and as a single cell protein. In addition to this, it is used as a hepatoprotectant and also acts as a lipotropic agent and prevents excess fat buildup in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Methionine is an essential amino acid required for tissue repair and maintenance

• L-Methionine affects absorption of selenium and zinc

• L-Methionine can assist cellular detoxification by chelating lead and mercury

• L-Methionine exhibitis properties associated with anti-aging and can also prevent the buildup of fat in the liver

Application

• L-Methionine is appropriate for use in cell culture and biochemistry laboratory applications

• It is useful for the laboratory study of methionine pathways in the cell relating to aging, degeneration, and metabolism

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
IUPAC Name (2S)-2-amino-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point ∼280°C (decomposition)
Beilstein 1722294
Merck Index 14,5975
Solubility Information Soluble in water and hydrochloride. Slightly soluble in acetic acid. Insoluble in ethanol,benzene,acetone and ether.
Quantity 500 g
Formula Weight 149.21
Optical Rotation +23° (c=2 in 2N HCl)
Percent Purity ≥98%
Chemical Name or Material L-Methionine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.