missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15214404 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
Product Code. Quantity Packaging unitSize
15214404 100 g Poly Bottle 100g
15567964 1 kg Poly Bottle 1kg
2 options
This item is not returnable. View return policy

Product Code. 15214404

Brand: Avantor J.T.Baker 208505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
SMILES CSCC[C@H](N)C(O)=O

Specifications

Packaging Poly Bottle
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Quantity 100 g
Formula Weight 149.21
Grade Multi-Compendial/USP
Chemical Name or Material L-Methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.