missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine, Pure, Fisher Chemical

Product Code. 10050280 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10050280 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10050280 Supplier Fisher Chemical Supplier No. M/4065/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 63-68-3
Color White
Molecular Formula C5H11NO2S
Packaging Plastic Powder Jar
MDL Number MFCD00063097 MFCD00801344
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
SMILES CSCC[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular Weight (g/mol) 149.21
Quantity 100 g
PubChem CID 6137
ChEBI CHEBI:16643
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.