missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine sulfone, 98%

Product Code. 10526830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10526830 5 g 5g
10371670 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10526830 Supplier Thermo Scientific Acros Supplier No. 439000050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7314-32-1 | C5H11NO4S | 181.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7314-32-1
Molecular Formula C5H11NO4S
Molecular Weight (g/mol) 181.21
MDL Number MFCD00066020
InChI Key UCUNFLYVYCGDHP-BYPYZUCNSA-N
Synonym l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+%
PubChem CID 445282
ChEBI CHEBI:21363
IUPAC Name (2S)-2-amino-4-methylsulfonylbutanoic acid
SMILES CS(=O)(=O)CC[C@H](N)C(O)=O

Specifications

Melting Point 242.3°C to 243.1°C
Color White
Elemental Analysis 32.31 to 33.97% C, 7.54 to 7.92% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Specific Rotation 12.7
Formula Weight 181.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Methionine sulfone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.