missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine Sulfoxide, TRC

Product Code. 30008213
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008213 5 g 5g
17927344 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008213 Supplier Toronto Research Chemicals Supplier No. TRCM2604505G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3226-65-1
Molecular Formula C5 H11 N O3 S
Molecular Weight (g/mol) 165.21
Synonym L-Methionine S-oxide, Butanoic acid, 2-amino-4-(methylsulfinyl)-, (2S)- (9CI), Butyric acid, 2-amino-4-(methylsulfinyl)-, L- (8CI), 3: PN: WO2011158590 PAGE: 34 claimed protein, L-Methionine sulfoxide, Methionine sulfoxide
IUPAC Name (2S)-2-amino-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H11NO3S/c1-10(9)3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-,10?/m0/s1
Quantity 5 g
Formula Weight 165.046
Percent Purity >95%
Shipping Condition Blue Ice
Chemical Name or Material L-Methionine Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.