missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine sulfoxide

Product Code. 15425179
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425179 25 g 25g
15405179 1 g 1g
15415179 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15425179 Supplier Thermo Scientific Chemicals Supplier No. J62873.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3226-65-1 | C5H11NO3S | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3226-65-1
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00063093
InChI Key QEFRNWWLZKMPFJ-YGVKFDHGSA-N
Synonym l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
PubChem CID 158980
ChEBI CHEBI:17016
IUPAC Name (2S)-2-amino-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](N)C(O)=O

Specifications

Color White
Beilstein 4129939
Quantity 25 g
Formula Weight 165.21
Physical Form Powder
Chemical Name or Material L-Methionine sulfoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.