missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine, TRC

Product Code. 30135079
Change view
Click to view available options
Quantity:
10 mg
1 g
10 g
Unit Size:
10g
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135079 10 g 10g
30010090 10 mg 10mg
30007181 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135079 Supplier Toronto Research Chemicals Supplier No. TRCM26044010G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5 H11 N O2 S
Molecular Weight (g/mol) 149.21
Synonym (2S)-2-Amino-4-(methylsulfanyl)butanoic acid, L-Methionine, Methionine, L- (8CI), (S)-2-Amino-4-(methylthio)butanoic acid, 2-Amino-4-(methylthio)butyric acid, Acimethin, Butanoic acid, 2-amino-4-(methylthio)-, (S)-, Cymethion, L-Homocysteine, S-methyl-, L-α-Amino-γ-methylthiobutyric acid, Meth-o-Tas, Methionine, Methiorep, NSC 22946, S-Methionine, S-Methyl-L-homocysteine, Secretory peptide (human clone HTXDU73), h-Met-oh, l-Methionine, α-Amino-γ-methylmercaptobutyric acid, γ-Methylthio-α-aminobutyric acid
IUPAC Name (2S)-2-amino-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1
Quantity 10 g
Formula Weight 149.051
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Methionine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.