missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10706074
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10706074 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10706074 Supplier Fisher BioReagents Supplier No. BP388100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 276°C
Ammonium 0.02% max.
CAS 63-68-3
Arsenic (As) 1ppm max.
pH 5.5 to 6.5
Color White
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Assay Percent Range 98.5 to 100.5 %
Loss on Drying 0.2% max.
Other Amino Acids Chromatographically not detectable
Molecular Formula C5H11NO2S
Packaging Glass Bottle
MDL Number MFCD00063097 MFCD00801344
Residue after Ignition 0.1% max.
Specific Rotation 23.0 to 24.5° (+ or -)
Merck Index 15, 6047
State of Solution (%T) 98.0% min.
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
Alpha Vector Methionine
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
SMILES CSCC[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-4-methylsulfanylbutanoic acid
Molecular Weight (g/mol) 149.21
Quantity 100 g
PubChem CID 6137
ChEBI CHEBI:16643
Formula Weight 149.21
Percent Purity 98.5 to 100.5%
Physical Form Solid
Chemical Name or Material L-Methionine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.