missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Norvaline, 99%

Product Code. 11392636
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392636 1 g 1g
11302646 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392636 Supplier Thermo Scientific Chemicals Supplier No. L08658.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6600-40-4 | C5H11NO2 | 117.15 g/mol

L-Norvaline enhances NO production from activated macrophages. It is known to promote tissue regeneration and muscle growth and become a precursor in the penicillin biosynthetic pathway. It also can be used for nutrition agent and drug synthesis. For example: it can react with phthalic acid anhydride to get N,N-phthaloyl-L-norvaline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Norvaline enhances NO production from activated macrophages. It is known to promote tissue regeneration and muscle growth and become a precursor in the penicillin biosynthetic pathway. It also can be used for nutrition agent and drug synthesis. For example: it can react with phthalic acid anhydride to get N,N-phthaloyl-L-norvaline.

Solubility
Soluble in water (almost transparency), 10.5 g/100 mL (18°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6600-40-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064421
InChI Key SNDPXSYFESPGGJ-BYPYZUCNSA-N
Synonym l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
PubChem CID 65098
ChEBI CHEBI:18314
IUPAC Name (2S)-2-aminopentanoic acid
SMILES CCC[C@H](N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721162
Merck Index 14,6716
Solubility Information Soluble in water (almost transparency),10.5g/100 mL (18°C).
Quantity 1 g
Formula Weight 117.15
Optical Rotation +25° (c=10 in 20% HCl)
Percent Purity 99%
Chemical Name or Material L-Norvaline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.