missing translation for 'onlineSavingsMsg'
Learn More

L-Octanoylcarnitine, MedChemExpress

Product Code. 16447487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447487 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16447487 Supplier Medchem Express Supplier No. HY113161
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress L-Octanoylcarnitine is the physiologically active form of octanoylcarnitine.

L-Octanoylcarnitine is the physiologically active form of octanoylcarnitine.

Specifications

Chemical Name or Material L-Octanoylcarnitine
CAS 25243-95-2
Color White
Purity Grade Notes Research
Molecular Formula C15H29NO4
Quantity 5 mg
Solubility Information DMSO : 5 mg/mL (17.40 mM; ultrasonic and warming and heat to 60°C)
SMILES CCCCCCCC(O[C@H](CC([O-])=O)C[N+](C)(C)C)=O
Molecular Weight (g/mol) 287.4
Formula Weight 287.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.