missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Ornithine HCL

Product Code. 30018638
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018638 100 g 100g
30018640 500 g 500g
30018639 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018638 Supplier Melford Supplier No. O20040100.0
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Polyamine growth regulator.

Polyamine growth regulator.

Specifications

CAS 3184-13-2
Molecular Formula C5H12N2O2-HCl
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.