missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Hydrochloride, 99%, MP Biomedicals

Product Code. 11471961 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
5 kg
Unit Size:
100g
25g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471961 25 g 25g
11481961 100 g 100g
11413100 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471961 Supplier MP Biomedicals Supplier No. 0219471825
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

L-Ornithine is a product of arginase degradation and plays an important role in ammonia metabolism via the urea cycle

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.621
MDL Number MFCD00064562
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Quantity 25 g
Formula Weight 168.6
Grade Cell Culture Reagent
Physical Form Crystalline
Chemical Name or Material L-Ornithine, 99%, Hydrochloride, This material is essentially free of citrulline and ammonia.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.