missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ornithine hydrochloride, 99%

Product Code. 10572141 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10572141

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3184-13-2 | C5H12N2O2·HCl | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H12N2O2·HCl
Molecular Weight (g/mol) 168.62
MDL Number MFCD00064562
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Melting Point 245.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula H2N(CH2)3CH(NH2)CO2H·HCl
Quantity 25 g
Beilstein 04, 420
Merck Index 14, 6874
Specific Rotation Condition + 24.00 (20.00°C c=4,6N HCl)
Specific Rotation + 24.00
Solubility Information (5% in 1 N HCl) Clear colorless
Formula Weight 168.62
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L(+)-Ornithine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.