missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Hydrochloride, TRC

Product Code. 30134077
Change view
Click to view available options
Quantity:
10 mg
5 g
25 g
Unit Size:
10mg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134077 25 g 25g
30010518 5 g 5g
30006654 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134077 Supplier Toronto Research Chemicals Supplier No. TRCO69555025G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5 H12 N2 O2 . Cl H
Molecular Weight (g/mol) 168.62
Synonym L-Ornithine, monohydrochloride (9CI), Ornithine, monohydrochloride, L- (8CI), L-(+)-Ornithine monohydrochloride, L-Ornithine hydrochloride, Ornithine hydrochloride, Ornithine monohydrochloride
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES Cl.NCCC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1
Quantity 25 g
Formula Weight 168.0666
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Ornithine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.