missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Perillaldehyde, 90%

Product Code. 10471821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10471821 25 g Glass bottle 25g
10461821 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10471821 Supplier Thermo Scientific Acros Supplier No. 203660250
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18031-40-8 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18031-40-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001543
InChI Key RUMOYJJNUMEFDD-SNVBAGLBSA-N
Synonym --perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s
PubChem CID 2724159
ChEBI CHEBI:86938
IUPAC Name (4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
SMILES CC(=C)C1CCC(=CC1)C=O

Specifications

Color Yellow
Density 0.9700g/mL
Boiling Point 104°C to 105°C (10.0 mmHg)
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 88% min
Packaging Glass bottle
Refractive Index 1.5040 to 1.51
Quantity 25 g
Beilstein 07, 158
Merck Index 14, 7167
Specific Gravity 0.97
Specific Rotation Condition − 115.00 (20.00°C c=10,C2H5OH)
Specific Rotation -115
Formula Weight 150.22
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material L(-)-Perillaldehyde, 90%
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Perillaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.