missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Phenylalanine, 99%

Product Code. 11478840
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1000 g
Unit Size:
1000g
100g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478840 100 g 100g
11468840 25 g 25g
11488840 500 g 500g
11400773 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11478840 Supplier Thermo Scientific Chemicals Supplier No. A13238.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-Phenylalanine, CAS # 63-91-2, is an essential amino acid with an aromatic structure used in the biosynthesis of other amino acids. It is important for the structure and function of many proteins and enzymes. | CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol

L-Phenylalanine is an aromatic organic compound containing amine and acid as functional groups. It is an essential amino acid and its structure can be considered as the derivative of 3-phenylpropionic acid, in which one hydrogen atom present at C-2 position is substituted with amino group. It is a chiral compound and consists of two optical isomers. It is also known as 2-amino-3-phenylpropanoic acid, beta-phenylalanine, L-alfa-amino-beta-phenylpropionic acid, H-Phe-OH and (S)-2-amino-3-phenylpropionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Phenylalanine is an essential amino acid found in several dietary sources, including meat, fish, eggs, cheese, and milk
  • Phenylalanine can be converted into the amino acid tyrosine

Application

  • L-Phenylalanine is used in the synthesis of other amino acids
  • It can be used as a precursor for the skin pigment melanin as well as the monoamine neurotransmitters dopamine, norepinephrine (noradrenaline), and epinephrine (adrenaline)
  • It is used to biochemically form proteins coded by DNA
  • Due to its ability to absorb ultraviolet radiation, it is used to quantify the amount of protein
  • In research, it is used as an alkaline phosphatase inhibitor
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064227
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point ∼275°C (decomposition)
Assay Percent Range 99%
Beilstein 1910408
Merck Index 14,7271
Solubility Information Soluble in water. Slightly soluble in methanol and ethanol. Insoluble in ethyl ether and benzene.
IUPAC Name (2S)-2-amino-3-phenylpropanoic acid
Quantity 100 g
Formula Weight 165.19
Optical Rotation −34° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Phenylalanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.