missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine ethyl ester hydrochloride, 98%

Product Code. p-7021330
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420184 5 g 5g
11430184 25 g 25g
11440184 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420184 Supplier Thermo Scientific Alfa Aesar Supplier No. A13618.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3182-93-2 | C11H16ClNO2 | 229.70 g/mol

L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Phenylalanine ethyl ester hydrochloride is used as an enzyme substrate.

Solubility
Soluble in methanol and water.

Notes
Keep container tightly sealed. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3182-93-2
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.70
MDL Number MFCD00012507
InChI Key FPFQPLFYTKMCHN-PPHPATTJSA-N
Synonym l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
PubChem CID 165085
IUPAC Name ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1

Specifications

Melting Point 153°C to 157°C
Beilstein 3657823
Solubility Information Soluble in methanol and water.
Quantity 5 g
Formula Weight 229.71
Optical Rotation +36° (c=5 in Ethanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material L-Phenylalanine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.