missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, Cell Culture Reagent, ≥99%, MP Biomedicals™

Product Code. 11421991 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421991 1 kg 1kg
11401991 25 g 25g
11411991 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421991 Supplier MP Biomedicals Supplier No. 0219472891
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-Proline, Cell Culture Reagent, ≥99% is used in cell culture media and as a component of MEM non-essential amino acids solution.

  • L-Proline is used in cell culture media.
  • L-Proline is also a component of MEM non-essential amino acids solution.

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
PubChem CID 145742
ChEBI CHEBI:17203
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

Melting Point 221°C
Solubility Information Soluble in water, 162,000 mg/L at 25°C
Density 1.064 g/mL at 24°C
Quantity 1 kg
Formula Weight 115.1
Optical Rotation -82° to -85° (c=1.6, water)
Percent Purity ≥99%
Grade Cell Culture
Physical Form Powder
pKa 1.95 (COOH), 10.64 (NH2)
Chemical Name or Material L-Proline, Hydroxy-L-proline free
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.