missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, 99%, MP Biomedicals, MP Biomedicals

Product Code. 11486082 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486082 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11486082 Supplier MP Biomedicals Supplier No. 0210273025
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 147-85-3
Molecular Formula C5H9NO2
MDL Number MFCD00064318
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
SMILES OC(=O)C1CCCN1
IUPAC Name pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 115.13
Quantity 25 g
PubChem CID 145742
ChEBI CHEBI:17203
Formula Weight 115.13
Physical Form Crystalline
Chemical Name or Material L-Proline, 99%, Hydroxy-L-proline Free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.