missing translation for 'onlineSavingsMsg'
Learn More

L-Proline methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11334176
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334176 1 g 1g
11344176 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334176 Supplier Thermo Scientific Alfa Aesar Supplier No. L19290.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2133-40-6
Molecular Formula C6H12NO2
Molecular Weight (g/mol) 130.17
MDL Number MFCD00012708
InChI Key BLWYXBNNBYXPPL-YFKPBYRVSA-O
Synonym l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride
PubChem CID 2733200
SMILES COC(=O)[C@@H]1CCC[NH2+]1

Specifications

Melting Point 71°C to 78°C
Beilstein 3596045
IUPAC Name methyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
Quantity 1 g
Formula Weight 165.62
Optical Rotation −33° (c=1 in Water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Proline methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.