missing translation for 'onlineSavingsMsg'
Learn More

L-Proline Methyl Ester Hydrochloride, TRC

Product Code. 30007091
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007091 1 g 1g
30004904 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007091 Supplier Toronto Research Chemicals Supplier No. TRCP7559801G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2133-40-6
Molecular Formula C6 H11 N O2 . Cl H
Molecular Weight (g/mol) 165.618
Synonym L-Proline, methyl ester, hydrochloride (1:1), L-Proline, methyl ester, hydrochloride (9CI), Proline, methyl ester, hydrochloride, L- (6CI,7CI,8CI), (2S)-(Methoxycarbonyl)pyrrolidine hydrochloride, (S)-Methyl pyrrolidine-2-carboxylate hydrochloride, (S)-Proline methyl ester hydrochloride, (S)-Pyrrolidine-2-carboxylic acid methyl ester hydrochloride, L-Pro-OMe hydrochloride, L-Proline methyl ester monohydrochloride, Methyl L-prolinate hydrochloride, Methyl L-prolinate hydrochloride, Methyl prolinate hydrochloride, Methyl proline hydrochloride, L-Proline methyl ester hydrochloride
IUPAC Name methyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES Cl.COC(=O)[C@@H]1CCCN1

Specifications

InChI Formula InChI=1S/C6H11NO2.ClH/c1-9-6(8)5-3-2-4-7-5;/h5,7H,2-4H2,1H3;1H/t5-;/m0./s1
Quantity 1 g
Formula Weight 165.0557
Shipping Condition Room Temperature
Chemical Name or Material L-Proline Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.