missing translation for 'onlineSavingsMsg'
Learn More

L-Proline methyl ester hydrochloride, 98%

Product Code. 10582271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10582271 5 g Glass Bottle 5g
10291333 1 g Glass Bottle 1g
10006393 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10582271 Supplier Thermo Scientific Acros Supplier No. 303000050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2133-40-6 | C6H12NO2 | 130.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2133-40-6
Molecular Formula C6H12NO2
Molecular Weight (g/mol) 130.17
MDL Number MFCD00012708
InChI Key BLWYXBNNBYXPPL-YFKPBYRVSA-O
Synonym l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride
PubChem CID 2733200
IUPAC Name methyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES COC(=O)[C@@H]1CCC[NH2+]1

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation -31° to -35° (20°C, 589nm) (c=1, H2O)
Water 0.5% max. (K.F.)
Specific Rotation Condition −33° (20°C c=1,H2O)
Quantity 5 g
Formula Weight 165.62
Percent Purity ≥97.5%
Physical Form Crystalline Powder, Crystals, and/or Chunks
Chemical Name or Material L-Proline methyleester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.