missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-butyl ester, 97%

Product Code. 10703902 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10703902 5 g Glass bottle 5g
10097829 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10703902 Supplier Thermo Scientific Acros Supplier No. 438110050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2812-46-6 | C9H17NO2 | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2812-46-6
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
InChI Key XJJBXZIKXFOMLP-ZETCQYMHSA-N
Synonym l-proline t-butyl ester, h-pro-otbu, tert-butyl l-prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, l-proline tert-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, l-proline t-butyl ester free base, t-butyl prolinate
PubChem CID 6453108
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate
SMILES CC(C)(C)OC(=O)C1CCCN1

Specifications

Color Colorless to Yellow
Boiling Point 50°C to 51°C (1.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation -40° to -48° (20°C, 589nm) (c=1, ethanol)
Formula Weight 171.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material L-Proline tert-butyl ester
Show More Show Less
compliance-icons
Product Identifier
  • L-Proline tert-butyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.