missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-butyl ester, 98%

Product Code. 15410469
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410469 5 g 5g
15400469 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410469 Supplier Thermo Scientific Alfa Aesar Supplier No. H63657.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2812-46-6 | C9H17NO2 | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2812-46-6
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00037879
InChI Key XJJBXZIKXFOMLP-ZETCQYMHSA-N
Synonym l-proline t-butyl ester, h-pro-otbu, tert-butyl l-prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, l-proline tert-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, l-proline t-butyl ester free base, t-butyl prolinate
PubChem CID 6453108
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate
SMILES CC(C)(C)OC(=O)C1CCCN1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 171.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material L-Proline tert-butyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.