missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Pyroglutamic Acid-13C5, TRC
Specifications
Specifications
| Chemical Name or Material | L-Pyroglutamic Acid-13C5 |
| CAS | 55443-56-6 |
| Molecular Formula | 13C5H7NO3 |
| Quantity | 5 mg |
| Synonym | (-)-2-Pyrrolidone-5-carboxylic Acid-13C5, (-)-Pyroglutamic Acid-13C5, (2S)-5-Oxopyrrolidine-2-carboxylic Acid-13C5, (5S)-2-Oxopyrrolidine-5-carboxylic Acid-13C5, (S)-(-)-2-Pyrrolidone-5-carboxylic Acid-13C5, (S)-(-)-γ-Butyrolactam-γ-carboxylic Acid-13C5, (S)-2-Pyrrolidone-5-carboxylic Acid-13C5, (S)-5-Oxo-2-pyrrolidinecarboxylic Acid-13C5, (S)-5-Oxoproline-13C5, (S)-Pyroglutamic Acid-13C5, 2-L-Pyrrolidone-5-carboxylic Acid-13C5, 2-Oxopyrrolidine-5(S)-carboxylic Acid, -13C5 2-Pyrrolidinone-5-carboxylic Acid-13C5, 5-Carboxy-2-pyrrolidinone-13C5, 5-Oxo-L-proline-13C5, 5-Oxoproline-13C5, 5-Pyrrolidinone-2-carboxylic Acid-13C5, Ajidew A 100-13C5, Glutimic Acid-13C5, Glutiminic Acid-13C5, L-(-)-Pyroglutamic Acid-13C5, L-2-Pyrrolidone-5-carboxylic Acid-13C5, L-5-Carboxy-2-pyrrolidinone-13C5, L-5-Oxo-2-pyrrolidinecarboxylic Acid-13C5, L-5-Oxoproline-13C5, γ-Lactam L-Glutamic Acid-13C5, L-Glutimic Acid-13C5, L-Glutiminic Acid, -13C5 L-Pyrrolidinonecarboxylic Acid-13C5, L-Pyrrolidonecarboxylic Acid-13C5, NSC 143034-13C5, Oxoproline-13C5, Oxopyrrolidinecarboxylic Acid-13C5, PCA-13C5, PCA 50-13C5, Pidolic Acid-13C5, Pidolidone-13C5, Pyroglutamic Acid-13C5, Pyrrolidinonecarboxylic Acid-13C5, Pyrrolidone-5-carboxylic Acid-13C5, Pyrrolidonecarboxylic Acid-13C5 |
| InChI Formula | InChI=1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1/i1+1,2+1,3+1,4+1,5+1 |
| SMILES | O=[13C]1[13CH2][13CH2][13C@]([H])(N1)[13C](O)=O |
| IUPAC Name | (S)-5-oxopyrrolidine-2-carboxylic-2,3,4,5-13C4 acid-13C |
| Molecular Weight (g/mol) | 134.08 |
| Formula Weight | 134.06 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?