missing translation for 'onlineSavingsMsg'
Learn More

L-Pyroglutamic Acid, TRC

Product Code. 30137168
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137168 100 g 100g
30135877 25 g 25g
30012233 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137168 Supplier Toronto Research Chemicals Supplier No. TRCP997480100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-79-3
Molecular Formula C5 H7 N O3
Molecular Weight (g/mol) 129.11
Synonym 5-Oxo-L-proline, Proline, 5-oxo-, L- (8CI), (-)-2-Pyrrolidone-5-carboxylic acid, (-)-Pyroglutamic acid, (2S)-5-Oxopyrrolidine-2-carboxylic acid, (5S)-2-Oxopyrrolidine-5-carboxylic acid, (S)-(-)-2-Pyrrolidone-5-carboxylic acid, (S)-(-)-γ-Butyrolactam-γ-carboxylic acid, (S)-2-Pyrrolidone-5-carboxylic acid, (S)-5-Oxo-1H-pyrrolidine-2-carboxylic acid, (S)-5-Oxo-2-pyrrolidinecarboxylic acid, (S)-5-Oxoproline, (S)-Pyroglutamic acid, 2-L-Pyrrolidone-5-carboxylic acid, 2-Oxopyrrolidine-5(S)-carboxylic acid, 2-Pyrrolidinone-5-carboxylic acid, 5-Carboxy-2-pyrrolidinone, 5-Oxo-L-proline, 5-Oxoproline, 5-Pyrrolidinone-2-carboxylic acid, Ajidew A 100, Glutimic acid, Glutiminic acid, L-(-)-Pyroglutamic acid, L-2-Pyrrolidone-5-carboxylic acid, L-5-Carboxy-2-pyrrolidinone, L-5-Oxo-2-pyrrolidinecarboxylic acid, L-5-Oxoproline, L-Glutamic acid, γ-lactam, L-Glutimic acid, L-Glutiminic acid, L-Pyroglutamic acid, L-Pyrrolidinonecarboxylic acid, L-Pyrrolidonecarboxylic acid, NSC 143034, Oxoproline, Oxopyrrolidinecarboxylic acid, PCA, PCA 50, Pidolic acid, Pidolidone, Pyroglutamic acid, Pyrrolidinonecarboxylic acid, Pyrrolidone-5-carboxylic acid, Pyrrolidonecarboxylic acid
IUPAC Name (2S)-5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)[C@@H]1CCC(=O)N1

Specifications

InChI Formula InChI=1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1
Quantity 100 g
Formula Weight 129.0426
Shipping Condition Room Temperature
Chemical Name or Material Pidolic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.