missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Rhamnose monohydrate, 99%

Product Code. 10583731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10583731 25 g Glass bottle 25g
10604852 100 g Glass bottle 100g
10464785 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10583731 Supplier Thermo Scientific Chemicals Supplier No. 174080250
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 10030-85-0 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10030-85-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00149363,MFCD00136036
InChI Key SHZGCJCMOBCMKK-HGVZOGFYSA-N
Synonym l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
PubChem CID 20849066
IUPAC Name (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate
SMILES C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 91.0°C to 93.0°C
Color White
Ignition Residue 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Specific Rotation Condition + 8.20 (20.00°C c=4.5,H2O)
Specific Rotation + 8.20
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in ethanol and methanol
Formula Weight 182.17
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Rhamnose monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.